Gene Transcripts
- Gene Symbol
- 2310002D06Rik
- Entrez Gene ID
- 69522
- Gene Name
- RIKEN cDNA 2310002D06 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 80507206 - 80517954 on Build GRCm38
- UniGene ID
- Mm.390530
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045490.1 |
|
2, 3 |
121 |
|
GenBank mRNA |
AK009088.1 |
- |
2, 3 |
122 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY708936.1 |
- |
2, 3 |
122 |
|
Chr. 12 - 80507206 - 80517904 [+] |
View Full Product Details