Gene Transcripts
- Gene Symbol
- 5430435K18Rik
- Entrez Gene ID
- 71357
- Gene Name
- RIKEN cDNA 5430435K18 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 151252144 - 151261400 on Build GRCm38
- UniGene ID
- Mm.389719
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017398.1 |
- |
1 |
157 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY717213.1 |
- |
1 |
157 |
|
Chr. 5 - 151252305 - 151259264 [+] |
View Full Product Details