Gene Transcripts
- Gene Symbol
- Gm16845
- Entrez Gene ID
- 100503652
- Gene Name
- predicted gene, 16845
- Gene Aliases
- Chromosome Location
- Chr. 9: 22071002 - 22086131 on Build GRCm38
- UniGene ID
- Mm.439941
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040406.1 |
|
4 |
851 |
|
NR_040407.1 |
|
3 |
787 |
|
GenBank mRNA |
AK154427.1 |
- |
3 |
757 |
|
View Full Product Details