Gene Transcripts
- Gene Symbol
- 1700010B09Rik
- Entrez Gene ID
- 69341
- Gene Name
- RIKEN cDNA 1700010B09 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.395378
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005812.1 |
- |
1 |
690 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY705965.1 |
- |
|
689 |
|
Chr. - - [-] |
View Full Product Details