Gene Transcripts
- Gene Symbol
- 7530416G11Rik
- Entrez Gene ID
- 328577
- Gene Name
- RIKEN cDNA 7530416G11 gene
- Gene Aliases
- Chromosome Location
- Chr. 15: 85472646 - 85503227 on Build GRCm38
- UniGene ID
- Mm.212960
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078701.1 |
- |
5 |
546 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB624027.1 |
- |
5 |
541 |
|
Chr. 15 - 85493239 - 85503223 [-] |
View Full Product Details