Gene Transcripts
- Gene Symbol
- LOC150935
- Entrez Gene ID
- 150935
- Gene Name
- uncharacterized LOC150935
- Gene Aliases
- Chromosome Location
- Chr. 2: 239762860 - 239800664 on Build GRCh38
- UniGene ID
- Hs.555582
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037808.1 |
|
3 |
580 |
|
GenBank mRNA |
AK056439.1 |
- |
3 |
580 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000407524.1 |
- |
3 |
580 |
|
Chr. 2 - 239762860 - 239802883 [+] |
View Full Product Details