Gene Transcripts
- Gene Symbol
- LOC101929378
- Entrez Gene ID
- 101929378
- Gene Name
- uncharacterized LOC101929378
- Gene Aliases
- Chromosome Location
- Chr. 2: 156020535 - 156254931 on Build GRCh38
- UniGene ID
- Hs.171192
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110249.2 |
|
4 |
1187 |
|
NR_110250.2 |
|
4 |
1237 |
|
GenBank mRNA |
BC032407.1 |
- |
4 |
1176 |
|
View Full Product Details