Gene Transcripts
- Gene Symbol
- LOC101927136
- Entrez Gene ID
- 101927136
- Gene Name
- uncharacterized LOC101927136
- Gene Aliases
- Chromosome Location
- Chr. 6: 53561289 - 53573275 on Build GRCh38
- UniGene ID
- Hs.509788
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000502390.5 |
- |
5 |
872 |
|
Chr. 6 - 53561289 - 53617007 [-] |
GENCODE |
ENST00000503985.5 |
- |
4 |
960 |
|
Chr. 6 - 53561906 - 53617171 [-] |
GENCODE |
ENST00000505995.5 |
- |
4 |
726 |
|
Chr. 6 - 53561920 - 53616994 [-] |
View Full Product Details