Gene Transcripts
- Gene Symbol
- Sox5it
- Entrez Gene ID
- 100038723
- Gene Name
- SRY (sex determining region Y)-box 5, intronic transcript
- Gene Aliases
- ENSMUSG00000072677, Gm10396
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.388565
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016013.1 |
- |
2 |
1106 |
|
View Full Product Details