Gene Transcripts
- Gene Symbol
- Gm4285
- Entrez Gene ID
- 100043189
- Gene Name
- predicted gene 4285
- Gene Aliases
- Chromosome Location
- Chr. 14: 75842918 - 75844964 on Build GRCm38
- UniGene ID
- Mm.426329
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045294.1 |
|
3 |
824 |
|
View Full Product Details