Gene Transcripts
- Gene Symbol
- LOC101927087
- Entrez Gene ID
- 101927087
- Gene Name
- uncharacterized LOC101927087
- Gene Aliases
- Chromosome Location
- Chr. 4: 124499942 - 124558434 on Build GRCh38
- UniGene ID
- Hs.518966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110838.1 |
|
9 |
783 |
|
GenBank mRNA |
AK057455.1 |
- |
9 |
783 |
|
View Full Product Details