Gene Transcripts

Gene Symbol
CDKL3
Entrez Gene ID
51265
Gene Name
cyclin dependent kinase like 3
Gene Aliases
NKIAMRE
Chromosome Location
Chr. 5: 134298424 - 134371159 on Build GRCh38
UniGene ID
Hs.719926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011543447.2 XP_011541749.1 2, 3 197
XM_017009547.1 XP_016865036.1 2, 3 234
GenBank mRNA AK297586.1 - 2, 3 164

View Full Product Details