Gene Transcripts

Gene Symbol
PLA2G1B
Entrez Gene ID
5319
Gene Name
phospholipase A2 group IB
Gene Aliases
PLA2, PLA2A, PPLA2
Chromosome Location
Chr. 12: 120322111 - 120327789 on Build GRCh38
UniGene ID
Hs.992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000928.2 NP_000919.1 4 434
GenBank mRNA AK311830.1 - 4 419
BC005386.1 - 3 296
BC106725.1 - 4 434 40014551
BC106726.1 - 4 434 40014552
BE969737.1 - 404
BX113838.1 - 3 238
CA867923.1 - 3 277
M21054.1 - 4 435

View Full Product Details