Gene Transcripts

Gene Symbol
Olfr610
Entrez Gene ID
259085
Gene Name
olfactory receptor 610
Gene Aliases
MOR9-2
Chromosome Location
Chr. 7: 103505997 - 103506944 on Build GRCm38
UniGene ID
Mm.377830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147081.2 NP_667292.2 1 694
GenBank mRNA BC113199.1 - 1 693

View Full Product Details