Gene Transcripts

Gene Symbol
Pon3
Entrez Gene ID
269823
Gene Name
paraoxonase 3
Gene Aliases
2810004E20, AI786302
Chromosome Location
Chr. 6: 5220852 - 5256250 on Build GRCm38
UniGene ID
Mm.9122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173006.1 NP_766594.1 4 365
GenBank mRNA AK076090.1 - 4 596
AK088572.1 - 4 365
BC099416.1 - 4 385 30847745
L76193.1 - 4 358

View Full Product Details