Gene Transcripts
- Gene Symbol
- LOC664787
- Entrez Gene ID
- 664787
- Gene Name
- Sp110 nuclear body protein pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 1|NT_162750: 17590 - 23437 on Build GRCm38
- UniGene ID
- Mm.393891
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087562.1 |
- |
7 |
1006 |
|
View Full Product Details