Gene Transcripts

Gene Symbol
Trmt61b
Entrez Gene ID
68789
Gene Name
tRNA methyltransferase 61B
Gene Aliases
1110039F03Rik, 623451, Gm6431
Chromosome Location
Chr. 17: 71557027 - 71598761 on Build GRCm38
UniGene ID
Mm.75202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AA268702.1 - 2, 3 121
AK004173.1 - 2, 3 353
AK032413.1 - 2, 3 846
AK036113.1 - 2, 3 855
AK045842.1 - 2, 3 807

View Full Product Details