Gene Transcripts

Gene Symbol
Acox1
Entrez Gene ID
50681
Gene Name
acyl-CoA oxidase 1, palmitoyl
Gene Aliases
RATACOA1
Chromosome Location
Chr. 10: 104724534 - 104748003 on Build Rnor_6.0
UniGene ID
Rn.31796
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017340.2 NP_059036.1 2029
GenBank mRNA BC085743.1 - 2029
J02752.1 - 2012

View Full Product Details