Gene Transcripts

Gene Symbol
Barhl2
Entrez Gene ID
65050
Gene Name
BarH-like homeobox 2
Gene Aliases
Barh, MBH1
Chromosome Location
Chr. 14: 4362717 - 4367614 on Build Rnor_6.0
UniGene ID
Rn.30005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022956.2 NP_075245.1 2 705
GenBank mRNA AB004056.1 - 2 709

View Full Product Details