Gene Transcripts
- Gene Symbol
- SSX3
- Entrez Gene ID
- 10214
- Gene Name
- SSX family member 3
- Gene Aliases
- CT5.3
- Chromosome Location
- Chr. X: 48346428 - 48356753 on Build GRCh38
- UniGene ID
- Hs.558445
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021014.3 |
NP_066294.1
|
8 |
995 |
|
GenBank mRNA |
U90840.1 |
- |
8 |
908 |
|
- Gene Symbol
- SSX2
- Entrez Gene ID
- 6757
- Gene Name
- SSX family member 2
- Gene Aliases
- CT5.2, CT5.2A, HD21, HOM-MEL-40, SSX
- Chromosome Location
- Chr. X: 52696896 - 52707227 on Build GRCh38
- UniGene ID
- Hs.289105
- Species
- Homo sapiens
- Gene Symbol
- SSX2B
- Entrez Gene ID
- 727837
- Gene Name
- SSX family member 2B
- Gene Aliases
- CT5.2, CT5.2b, HOM-MEL-40, SSX
- Chromosome Location
- Chr. X: 52751204 - 52761536 on Build GRCh38
- UniGene ID
- Hs.289105
- Species
- Homo sapiens
View Full Product Details