Gene Transcripts

Gene Symbol
Clta
Entrez Gene ID
83800
Gene Name
clathrin, light chain A
Gene Aliases
LCA1, LCA2, LCA3
Chromosome Location
Chr. 5: 59490363 - 59509139 on Build Rnor_6.0
UniGene ID
Rn.112599
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031974.1 NP_114180.1 6 675
XM_006238105.1 XP_006238167.1 5 1394
GenBank mRNA FQ213034.1 - 6 683
M15882.1 - 6 675

View Full Product Details