Gene Transcripts

Gene Symbol
LOC102551847
Entrez Gene ID
102551847
Gene Name
uncharacterized LOC102551847
Gene Aliases
Chromosome Location
Chr. X: 68474557 - 68481462 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773300.1 XP_008771522.1 1 663

View Full Product Details