Gene Transcripts

Gene Symbol
Orai1
Entrez Gene ID
304496
Gene Name
ORAI calcium release-activated calcium modulator 1
Gene Aliases
RGD1311873
Chromosome Location
Chr. 12: 38981903 - 38995570 on Build Rnor_6.0
UniGene ID
Rn.3869
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013982.1 NP_001014004.1 2 862
GenBank mRNA BC088225.1 - 2 862
EU644749.1 - 2 777

View Full Product Details