Gene Transcripts

Gene Symbol
ARSB
Entrez Gene ID
411
Gene Name
arylsulfatase B
Gene Aliases
ASB, G4S, MPS6
Chromosome Location
Chr. 5: 78777209 - 78986534 on Build GRCh38
UniGene ID
Hs.149103
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000046.3 NP_000037.2 2 1616
NM_198709.2 NP_942002.1 3 685
XM_011543390.1 XP_011541692.1 3 829
XM_011543391.2 XP_011541693.1 2 392
XM_011543392.2 XP_011541694.1 2 392
XM_011543393.1 XP_011541695.1 2 392
XM_017009471.1 XP_016864960.1 2 392
GenBank mRNA AK290865.1 - 2 392
BC029051.1 - 3 685 5186657
J05225.1 - 2 336
M32373.1 - 2 889

View Full Product Details