Gene Transcripts

Gene Symbol
Stard3
Entrez Gene ID
363675
Gene Name
StAR-related lipid transfer (START) domain containing 3
Gene Aliases
Chromosome Location
Chr. 10: 86313430 - 86335864 on Build Rnor_6.0
UniGene ID
Rn.145430
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014229.1 NP_001014251.1 11 1038
GenBank mRNA BC085872.1 - 11 1038

View Full Product Details