Gene Transcripts

Gene Symbol
Zc3h13
Entrez Gene ID
305955
Gene Name
zinc finger CCCH type containing 13
Gene Aliases
RGD1307729
Chromosome Location
Chr. 15: 57340519 - 57405287 on Build Rnor_6.0
UniGene ID
Rn.100827
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170471.1 NP_001163942.1 18 5287
XM_006252316.2 XP_006252378.1 18 5343
XM_006252317.2 XP_006252379.1 18 5344

View Full Product Details