Gene Transcripts

Gene Symbol
Rhox8
Entrez Gene ID
503423
Gene Name
reproductive homeobox 8
Gene Aliases
Chromosome Location
Chr. X: 123976734 - 123980357 on Build Rnor_6.0
UniGene ID
Rn.121413
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025776.1 NP_001020947.1 1, 2 75
GenBank mRNA DQ058655.1 - 1, 2 75

View Full Product Details