Gene Transcripts

Gene Symbol
LOC686288
Entrez Gene ID
686288
Gene Name
similar to olfactory receptor Olr1668
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.142702
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101012.1 NP_001094482.1 4 964
GenBank mRNA BC128716.1 - 4 964

View Full Product Details