Gene Transcripts

Gene Symbol
Six3
Entrez Gene ID
20473
Gene Name
sine oculis-related homeobox 3
Gene Aliases
E130112M24Rik, Six3a, Six3alpha, Six3b, Six3beta
Chromosome Location
Chr. 17: 85620834 - 85626191 on Build GRCm38
UniGene ID
Mm.370208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011381.4 NP_035511.2 2 1501
GenBank mRNA BC098096.1 - 2 1121
CD352849.1 - 2 559
D83145.1 - 2 1087
X90871.1 - 1274

View Full Product Details