Gene Transcripts

Gene Symbol
Cldn3
Entrez Gene ID
65130
Gene Name
claudin 3
Gene Aliases
Chromosome Location
Chr. 12: 21831341 - 21832813 on Build Rnor_6.0
UniGene ID
Rn.4513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031700.2 NP_113888.2 1 561
GenBank mRNA BC062411.1 - 1 186
M74067.1 - 1 284

View Full Product Details