Gene Transcripts

Gene Symbol
KIR2DL3
Entrez Gene ID
3804
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3
Gene Aliases
CD158B2, CD158b, GL183, KIR-023GB, KIR-K7b, KIR-K7c, KIR2DS3, KIR2DS5, KIRCL23, NKAT, NKAT2, NKAT2A, NKAT2B, p58
Chromosome Location
Chr. 19: 54738509 - 54753052 on Build GRCh38
UniGene ID
Hs.654608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015868.2 NP_056952.2 4, 5 693
GenBank mRNA AF022048.1 - 4, 5 656
AF135566.1 - 2 443
AF135567.1 - 2 482
AF285435.1 - 3, 4 616
AY789057.1 - 4, 5 656
BC032422.1 - 4, 5 669 5220339
BC050730.1 - 4, 5 687 5219336
BC110613.1 - 4, 5 670 40031619
FN395076.1 - 4, 5 656
FN552550.1 - 4, 5 638
L41268.1 - 4, 5 693
L76662.1 - 4, 5 667
L76663.1 - 4, 5 667
U24074.1 - 4, 5 677
U50546.1 - 4, 5 373
U73395.1 - 4, 5 677
U96190.1 - 2 593

Gene Symbol
KIR2DL1
Entrez Gene ID
3802
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1
Gene Aliases
CD158A, KIR-K64, KIR221, KIR2DS1, NKAT, NKAT-1, NKAT1, p58.1
Chromosome Location
Chr. 19: 54769208 - 54784326 on Build GRCh38
UniGene ID
Hs.654605
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014218.2 NP_055033.2 4, 5 694
XM_017026782.1 XP_016882271.1 4, 5 672
XM_017026783.1 XP_016882272.1 4, 5 1299
GenBank mRNA AF022045.1 - 4, 5 663
AF135560.1 - 2 593
AF285431.1 - 4, 5 689
AF285432.1 - 3, 4 616
AY366237.1 - 4, 5 692
AY366238.1 - 4, 5 638
AY366239.1 - 4, 5 686
AY366240.1 - 4, 5 694
AY366241.1 - 4, 5 693
AY388471.1 - 4, 5 686
AY789055.1 - 5, 6 689
AY789056.1 - 4, 5 656
BC069344.1 - 4, 5 689 7262296
KJ699235.1 - 4, 5 929
L41267.1 - 5, 6 688
U24076.1 - 5, 6 689
U24078.1 - 4, 5 696
U96189.1 - 2 593

Gene Symbol
KIR2DL2
Entrez Gene ID
3803
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 2
Gene Aliases
CD158B1, CD158b, NKAT-6, NKAT6, p58.2
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.743949
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014219.2 NP_055034.2 4, 5 685
GenBank mRNA AF285434.1 - 3, 4 616

Gene Symbol
KIR2DL5A
Entrez Gene ID
57292
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5A
Gene Aliases
CD158F, KIR2DL5, KIR2DL5.1, KIR2DL5.3
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.740906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020535.3 NP_065396.1 4, 5 696
GenBank mRNA AF204903.1 - 4, 5 696
AF217487.1 - 6, 7 665

Gene Symbol
KIR2DL5B
Entrez Gene ID
553128
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5B
Gene Aliases
KIR2DL5, KIR2DL5.2, KIR2DLX
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.740906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018081.1 NP_001018091.1 4, 5 641
GenBank mRNA AF271607.1 - 4, 5 641
AF271608.1 - 4, 5 641
AJ544587.1 - 5, 6 642

Gene Symbol
KIR2DS1
Entrez Gene ID
3806
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1
Gene Aliases
CD158H, CD158a, KIR2DP1DL1, KIR2DS4, p50.1
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.744319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014512.1 NP_055327.1 4, 5 669
GenBank mRNA AF022046.1 - 4, 5 669
AF285437.1 - 3, 4 616
AM999888.1 - 4, 5 664
X89892.1 - 4, 5 676
X98858.1 - 4, 5 656

Gene Symbol
KIR2DS2
Entrez Gene ID
100132285
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2
Gene Aliases
183ActI, CD158J, CD158b, KIR-2DS2, NKAT-5, NKAT5, cl-49
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.512572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291695.1 NP_001278624.1 4, 5 691
NM_001291696.1 NP_001278625.1 3, 4 391
NM_012312.4 NP_036444.1 4, 5 691
GenBank mRNA AF135565.1 - 2 299
AF285438.1 - 3, 4 616
AJ002102.1 - 4, 5 653
AJ002103.1 - 4, 5 656
AM409094.1 - 4, 5 657
AY366243.1 - 4, 5 666
AY366244.1 - 4, 5 655
BC108917.1 - 4, 5 658 40017299
BC119631.1 - 4, 5 675 40115568
BC119632.1 - 4, 5 675 40115570
BC125146.1 - 3, 4 640
BC126856.1 - 4, 5 658 40017301
FR750994.1 - 4, 5 691
L41347.1 - 4, 5 675
L76667.1 - 4, 5 373
L76668.1 - 3, 4 367
U24079.1 - 4, 5 654
X89893.1 - 4, 5 656

Gene Symbol
KIR2DS3
Entrez Gene ID
3808
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 3
Gene Aliases
NKAT7
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.744266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012313.1 NP_036445.1 4, 5 667
GenBank mRNA AF022047.1 - 4, 5 660
AM851002.1 - 4, 5 692
AY789063.2 - 3, 4 640
L76670.1 - 4, 5 667
X97231.1 - 4, 5 715

Gene Symbol
KIR2DS4
Entrez Gene ID
3809
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
Gene Aliases
CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
Chromosome Location
Chr. 19: 54832676 - 54848569 on Build GRCh38
UniGene ID
Hs.654608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF135564.1 - 4 497
U96188.1 - 3 593

Gene Symbol
KIR2DS5
Entrez Gene ID
3810
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5
Gene Aliases
CD158G, NKAT9
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.744266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014513.2 NP_055328.2 4, 5 690
GenBank mRNA AF208054.1 - 4, 5 690
AF272389.1 - 5, 6 640
AY366247.1 - 4, 5 714
AY366248.1 - 6, 7 714
L76672.1 - 4, 5 667

Gene Symbol
KIR3DL1
Entrez Gene ID
3811
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
Gene Aliases
CD158E1, KIR, KIR3DL1/S1, KIR3DL2, NKAT-3, NKAT3, NKB1, NKB1B
Chromosome Location
Chr. 19: 54816438 - 54830778 on Build GRCh38
UniGene ID
Hs.645228
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322168.1 NP_001309097.1 5, 6 1004
NM_013289.2 NP_037421.2 5, 6 1004
GenBank mRNA AF022049.1 - 5, 6 941
AF103010.1 - 5, 6 941
AF103011.1 - 5, 6 941
AF103012.1 - 5, 6 941
AF103013.1 - 5, 6 941
AF208688.1 - 3 878
AF262968.1 - 3, 4 860
AF262969.1 - 3, 4 860
AF262970.1 - 3, 4 860
AF262971.1 - 3, 4 860
AF262972.1 - 3, 4 860
AF262973.1 - 5, 6 974
AF262974.1 - 5, 6 974
AJ417556.1 - 3, 4 890
AJ417557.1 - 3, 4 890
AY366236.1 - 5, 6 979
AY366249.1 - 5, 6 941
AY366250.1 - 5, 6 967
AY366251.1 - 5, 6 941
AY366252.1 - 5, 6 941
AY760050.1 - 5, 6 941
AY789065.1 - 5, 6 956
AY789066.1 - 5, 6 941
AY789072.1 - 5, 6 941
BC028206.1 - 5, 6 968 5215674
BC101540.1 - 5, 6 960 8069046
BC101542.1 - 5, 6 960 8069048
DQ178659.1 - 5, 6 941
EF462885.1 - 3, 4 871
EF472674.1 - 3, 4 871
EF472675.1 - 3, 4 871
EF472676.1 - 3, 4 871
EF472677.1 - 3, 4 871
EF472678.1 - 3, 4 871
EF472679.1 - 3, 4 871
L41269.1 - 5, 6 960
L76664.1 - 4, 5 667
U30273.1 - 5, 6 943
U30274.1 - 5, 6 1004
U31416.1 - 5, 6 966
U33328.1 - 5, 6 966
X94262.1 - 5, 6 942

Gene Symbol
KIR3DL2
Entrez Gene ID
3812
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2
Gene Aliases
3DL2, CD158K, KIR-3DL2, NKAT-4, NKAT4, NKAT4B, p140
Chromosome Location
Chr. 19: 54850319 - 54867215 on Build GRCh38
UniGene ID
Hs.645228
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006737.3 NP_006728.2 5, 6 974
XM_011526940.2 XP_011525242.1 5, 6 1098
XM_017026784.1 XP_016882273.1 5, 6 1098
GenBank mRNA AF208683.1 - 3 878
AF208684.1 - 3 878
AF208685.1 - 3 878
AF208686.1 - 3 878
AF208687.1 - 3 878
AF208690.1 - 3 791
AF262965.1 - 3, 4 860
AF262966.1 - 3, 4 860
AF262967.1 - 5, 6 974
AF263617.1 - 5, 6 974
AJ276125.1 - 5, 6 942
AY059418.1 - 3, 4 872
AY059419.1 - 3, 4 872
AY059420.1 - 3, 4 872
AY366256.2 - 5, 6 953
AY789067.1 - 5, 6 941
AY789068.1 - 5, 6 941
AY789069.1 - 4, 5 911
AY789070.1 - 5, 6 941
AY789071.1 - 4, 5 911
BC105678.1 - 5, 6 966 40004817
BC107951.1 - 5, 6 966 40004820
DQ256504.1 - 3, 4 873
EF582384.1 - 5, 6 941
L41270.1 - 5, 6 960
L76665.1 - 5, 6 952
L76666.1 - 5, 6 952
U30272.1 - 5, 6 943
X93595.1 - 5, 6 971
X94373.1 - 5, 6 942
X94374.1 - 5, 6 941

Gene Symbol
KIR3DS1
Entrez Gene ID
3813
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1
Gene Aliases
CD158E2, KIR-123FM, KIR-G1, NKAT-10, NKAT10
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.743496
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083539.2 NP_001077008.1 5, 6 978
NM_001282170.1 NP_001269099.1 4, 5 693
NM_001282171.1 NP_001269100.1 3, 4 657
GenBank mRNA AF022044.1 - 5, 6 954
AJ417558.3 - 3, 4 890
AK301140.1 - 5, 6 978
BC098107.1 - 3, 4 631 40026977
BC098288.1 - 5, 6 952 40026980
BC105679.1 - 5, 6 952 40004313
EF370406.1 - 4, 5 901
EF582383.1 - 5, 6 941
L76661.1 - 5, 6 952
U73396.1 - 5, 6 941
X97232.1 - 4, 5 656
X97233.1 - 5, 6 941

View Full Product Details