Gene Transcripts
- Gene Symbol
- KIR2DL3
- Entrez Gene ID
- 3804
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3
- Gene Aliases
- CD158B2, CD158b, GL183, KIR-023GB, KIR-K7b, KIR-K7c, KIR2DS3, KIR2DS5, KIRCL23, NKAT, NKAT2, NKAT2A, NKAT2B, p58
- Chromosome Location
- Chr. 19: 54738509 - 54753052 on Build GRCh38
- UniGene ID
- Hs.654608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015868.2 |
NP_056952.2
|
4, 5 |
693 |
|
GenBank mRNA |
AF022048.1 |
- |
4, 5 |
656 |
|
AF135566.1 |
- |
2 |
443 |
|
AF135567.1 |
- |
2 |
482 |
|
AF285435.1 |
- |
3, 4 |
616 |
|
AY789057.1 |
- |
4, 5 |
656 |
|
BC032422.1 |
- |
4, 5 |
669 |
5220339 |
BC050730.1 |
- |
4, 5 |
687 |
5219336 |
BC110613.1 |
- |
4, 5 |
670 |
40031619 |
FN395076.1 |
- |
4, 5 |
656 |
|
FN552550.1 |
- |
4, 5 |
638 |
|
L41268.1 |
- |
4, 5 |
693 |
|
L76662.1 |
- |
4, 5 |
667 |
|
L76663.1 |
- |
4, 5 |
667 |
|
U24074.1 |
- |
4, 5 |
677 |
|
U50546.1 |
- |
4, 5 |
373 |
|
U73395.1 |
- |
4, 5 |
677 |
|
U96190.1 |
- |
2 |
593 |
|
- Gene Symbol
- KIR2DL1
- Entrez Gene ID
- 3802
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1
- Gene Aliases
- CD158A, KIR-K64, KIR221, KIR2DS1, NKAT, NKAT-1, NKAT1, p58.1
- Chromosome Location
- Chr. 19: 54769208 - 54784326 on Build GRCh38
- UniGene ID
- Hs.654605
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014218.2 |
NP_055033.2
|
4, 5 |
694 |
|
XM_017026782.1 |
XP_016882271.1
|
4, 5 |
672 |
|
XM_017026783.1 |
XP_016882272.1
|
4, 5 |
1299 |
|
GenBank mRNA |
AF022045.1 |
- |
4, 5 |
663 |
|
AF135560.1 |
- |
2 |
593 |
|
AF285431.1 |
- |
4, 5 |
689 |
|
AF285432.1 |
- |
3, 4 |
616 |
|
AY366237.1 |
- |
4, 5 |
692 |
|
AY366238.1 |
- |
4, 5 |
638 |
|
AY366239.1 |
- |
4, 5 |
686 |
|
AY366240.1 |
- |
4, 5 |
694 |
|
AY366241.1 |
- |
4, 5 |
693 |
|
AY388471.1 |
- |
4, 5 |
686 |
|
AY789055.1 |
- |
5, 6 |
689 |
|
AY789056.1 |
- |
4, 5 |
656 |
|
BC069344.1 |
- |
4, 5 |
689 |
7262296 |
KJ699235.1 |
- |
4, 5 |
929 |
|
L41267.1 |
- |
5, 6 |
688 |
|
U24076.1 |
- |
5, 6 |
689 |
|
U24078.1 |
- |
4, 5 |
696 |
|
U96189.1 |
- |
2 |
593 |
|
- Gene Symbol
- KIR2DL2
- Entrez Gene ID
- 3803
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 2
- Gene Aliases
- CD158B1, CD158b, NKAT-6, NKAT6, p58.2
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.743949
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014219.2 |
NP_055034.2
|
4, 5 |
685 |
|
GenBank mRNA |
AF285434.1 |
- |
3, 4 |
616 |
|
- Gene Symbol
- KIR2DL5A
- Entrez Gene ID
- 57292
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5A
- Gene Aliases
- CD158F, KIR2DL5, KIR2DL5.1, KIR2DL5.3
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.740906
- Species
- Homo sapiens
- Gene Symbol
- KIR2DL5B
- Entrez Gene ID
- 553128
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5B
- Gene Aliases
- KIR2DL5, KIR2DL5.2, KIR2DLX
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.740906
- Species
- Homo sapiens
- Gene Symbol
- KIR2DS1
- Entrez Gene ID
- 3806
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1
- Gene Aliases
- CD158H, CD158a, KIR2DP1DL1, KIR2DS4, p50.1
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.744319
- Species
- Homo sapiens
- Gene Symbol
- KIR2DS2
- Entrez Gene ID
- 100132285
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2
- Gene Aliases
- 183ActI, CD158J, CD158b, KIR-2DS2, NKAT-5, NKAT5, cl-49
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.512572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291695.1 |
NP_001278624.1
|
4, 5 |
691 |
|
NM_001291696.1 |
NP_001278625.1
|
3, 4 |
391 |
|
NM_012312.4 |
NP_036444.1
|
4, 5 |
691 |
|
GenBank mRNA |
AF135565.1 |
- |
2 |
299 |
|
AF285438.1 |
- |
3, 4 |
616 |
|
AJ002102.1 |
- |
4, 5 |
653 |
|
AJ002103.1 |
- |
4, 5 |
656 |
|
AM409094.1 |
- |
4, 5 |
657 |
|
AY366243.1 |
- |
4, 5 |
666 |
|
AY366244.1 |
- |
4, 5 |
655 |
|
BC108917.1 |
- |
4, 5 |
658 |
40017299 |
BC119631.1 |
- |
4, 5 |
675 |
40115568 |
BC119632.1 |
- |
4, 5 |
675 |
40115570 |
BC125146.1 |
- |
3, 4 |
640 |
|
BC126856.1 |
- |
4, 5 |
658 |
40017301 |
FR750994.1 |
- |
4, 5 |
691 |
|
L41347.1 |
- |
4, 5 |
675 |
|
L76667.1 |
- |
4, 5 |
373 |
|
L76668.1 |
- |
3, 4 |
367 |
|
U24079.1 |
- |
4, 5 |
654 |
|
X89893.1 |
- |
4, 5 |
656 |
|
- Gene Symbol
- KIR2DS3
- Entrez Gene ID
- 3808
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 3
- Gene Aliases
- NKAT7
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.744266
- Species
- Homo sapiens
- Gene Symbol
- KIR2DS4
- Entrez Gene ID
- 3809
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
- Gene Aliases
- CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
- Chromosome Location
- Chr. 19: 54832676 - 54848569 on Build GRCh38
- UniGene ID
- Hs.654608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF135564.1 |
- |
4 |
497 |
|
U96188.1 |
- |
3 |
593 |
|
- Gene Symbol
- KIR2DS5
- Entrez Gene ID
- 3810
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5
- Gene Aliases
- CD158G, NKAT9
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.744266
- Species
- Homo sapiens
- Gene Symbol
- KIR3DL1
- Entrez Gene ID
- 3811
- Gene Name
- killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
- Gene Aliases
- CD158E1, KIR, KIR3DL1/S1, KIR3DL2, NKAT-3, NKAT3, NKB1, NKB1B
- Chromosome Location
- Chr. 19: 54816438 - 54830778 on Build GRCh38
- UniGene ID
- Hs.645228
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001322168.1 |
NP_001309097.1
|
5, 6 |
1004 |
|
NM_013289.2 |
NP_037421.2
|
5, 6 |
1004 |
|
GenBank mRNA |
AF022049.1 |
- |
5, 6 |
941 |
|
AF103010.1 |
- |
5, 6 |
941 |
|
AF103011.1 |
- |
5, 6 |
941 |
|
AF103012.1 |
- |
5, 6 |
941 |
|
AF103013.1 |
- |
5, 6 |
941 |
|
AF208688.1 |
- |
3 |
878 |
|
AF262968.1 |
- |
3, 4 |
860 |
|
AF262969.1 |
- |
3, 4 |
860 |
|
AF262970.1 |
- |
3, 4 |
860 |
|
AF262971.1 |
- |
3, 4 |
860 |
|
AF262972.1 |
- |
3, 4 |
860 |
|
AF262973.1 |
- |
5, 6 |
974 |
|
AF262974.1 |
- |
5, 6 |
974 |
|
AJ417556.1 |
- |
3, 4 |
890 |
|
AJ417557.1 |
- |
3, 4 |
890 |
|
AY366236.1 |
- |
5, 6 |
979 |
|
AY366249.1 |
- |
5, 6 |
941 |
|
AY366250.1 |
- |
5, 6 |
967 |
|
AY366251.1 |
- |
5, 6 |
941 |
|
AY366252.1 |
- |
5, 6 |
941 |
|
AY760050.1 |
- |
5, 6 |
941 |
|
AY789065.1 |
- |
5, 6 |
956 |
|
AY789066.1 |
- |
5, 6 |
941 |
|
AY789072.1 |
- |
5, 6 |
941 |
|
BC028206.1 |
- |
5, 6 |
968 |
5215674 |
BC101540.1 |
- |
5, 6 |
960 |
8069046 |
BC101542.1 |
- |
5, 6 |
960 |
8069048 |
DQ178659.1 |
- |
5, 6 |
941 |
|
EF462885.1 |
- |
3, 4 |
871 |
|
EF472674.1 |
- |
3, 4 |
871 |
|
EF472675.1 |
- |
3, 4 |
871 |
|
EF472676.1 |
- |
3, 4 |
871 |
|
EF472677.1 |
- |
3, 4 |
871 |
|
EF472678.1 |
- |
3, 4 |
871 |
|
EF472679.1 |
- |
3, 4 |
871 |
|
L41269.1 |
- |
5, 6 |
960 |
|
L76664.1 |
- |
4, 5 |
667 |
|
U30273.1 |
- |
5, 6 |
943 |
|
U30274.1 |
- |
5, 6 |
1004 |
|
U31416.1 |
- |
5, 6 |
966 |
|
U33328.1 |
- |
5, 6 |
966 |
|
X94262.1 |
- |
5, 6 |
942 |
|
- Gene Symbol
- KIR3DL2
- Entrez Gene ID
- 3812
- Gene Name
- killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2
- Gene Aliases
- 3DL2, CD158K, KIR-3DL2, NKAT-4, NKAT4, NKAT4B, p140
- Chromosome Location
- Chr. 19: 54850319 - 54867215 on Build GRCh38
- UniGene ID
- Hs.645228
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006737.3 |
NP_006728.2
|
5, 6 |
974 |
|
XM_011526940.2 |
XP_011525242.1
|
5, 6 |
1098 |
|
XM_017026784.1 |
XP_016882273.1
|
5, 6 |
1098 |
|
GenBank mRNA |
AF208683.1 |
- |
3 |
878 |
|
AF208684.1 |
- |
3 |
878 |
|
AF208685.1 |
- |
3 |
878 |
|
AF208686.1 |
- |
3 |
878 |
|
AF208687.1 |
- |
3 |
878 |
|
AF208690.1 |
- |
3 |
791 |
|
AF262965.1 |
- |
3, 4 |
860 |
|
AF262966.1 |
- |
3, 4 |
860 |
|
AF262967.1 |
- |
5, 6 |
974 |
|
AF263617.1 |
- |
5, 6 |
974 |
|
AJ276125.1 |
- |
5, 6 |
942 |
|
AY059418.1 |
- |
3, 4 |
872 |
|
AY059419.1 |
- |
3, 4 |
872 |
|
AY059420.1 |
- |
3, 4 |
872 |
|
AY366256.2 |
- |
5, 6 |
953 |
|
AY789067.1 |
- |
5, 6 |
941 |
|
AY789068.1 |
- |
5, 6 |
941 |
|
AY789069.1 |
- |
4, 5 |
911 |
|
AY789070.1 |
- |
5, 6 |
941 |
|
AY789071.1 |
- |
4, 5 |
911 |
|
BC105678.1 |
- |
5, 6 |
966 |
40004817 |
BC107951.1 |
- |
5, 6 |
966 |
40004820 |
DQ256504.1 |
- |
3, 4 |
873 |
|
EF582384.1 |
- |
5, 6 |
941 |
|
L41270.1 |
- |
5, 6 |
960 |
|
L76665.1 |
- |
5, 6 |
952 |
|
L76666.1 |
- |
5, 6 |
952 |
|
U30272.1 |
- |
5, 6 |
943 |
|
X93595.1 |
- |
5, 6 |
971 |
|
X94373.1 |
- |
5, 6 |
942 |
|
X94374.1 |
- |
5, 6 |
941 |
|
- Gene Symbol
- KIR3DS1
- Entrez Gene ID
- 3813
- Gene Name
- killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1
- Gene Aliases
- CD158E2, KIR-123FM, KIR-G1, NKAT-10, NKAT10
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.743496
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083539.2 |
NP_001077008.1
|
5, 6 |
978 |
|
NM_001282170.1 |
NP_001269099.1
|
4, 5 |
693 |
|
NM_001282171.1 |
NP_001269100.1
|
3, 4 |
657 |
|
GenBank mRNA |
AF022044.1 |
- |
5, 6 |
954 |
|
AJ417558.3 |
- |
3, 4 |
890 |
|
AK301140.1 |
- |
5, 6 |
978 |
|
BC098107.1 |
- |
3, 4 |
631 |
40026977 |
BC098288.1 |
- |
5, 6 |
952 |
40026980 |
BC105679.1 |
- |
5, 6 |
952 |
40004313 |
EF370406.1 |
- |
4, 5 |
901 |
|
EF582383.1 |
- |
5, 6 |
941 |
|
L76661.1 |
- |
5, 6 |
952 |
|
U73396.1 |
- |
5, 6 |
941 |
|
X97232.1 |
- |
4, 5 |
656 |
|
X97233.1 |
- |
5, 6 |
941 |
|
View Full Product Details