Gene Transcripts
- Gene Symbol
- SSX4
- Entrez Gene ID
- 6759
- Gene Name
- SSX family member 4
- Gene Aliases
- CT5.4
- Chromosome Location
- Chr. X: 48383527 - 48393343 on Build GRCh38
- UniGene ID
- Hs.558402
- Species
- Homo sapiens
- Gene Symbol
- SSX1
- Entrez Gene ID
- 6756
- Gene Name
- SSX family member 1
- Gene Aliases
- CT5.1, SSRC
- Chromosome Location
- Chr. X: 48255317 - 48267444 on Build GRCh38
- UniGene ID
- Hs.434142
- Species
- Homo sapiens
- Gene Symbol
- SSX2
- Entrez Gene ID
- 6757
- Gene Name
- SSX family member 2
- Gene Aliases
- CT5.2, CT5.2A, HD21, HOM-MEL-40, SSX
- Chromosome Location
- Chr. X: 52696896 - 52707227 on Build GRCh38
- UniGene ID
- Hs.289105
- Species
- Homo sapiens
- Gene Symbol
- SSX2B
- Entrez Gene ID
- 727837
- Gene Name
- SSX family member 2B
- Gene Aliases
- CT5.2, CT5.2b, HOM-MEL-40, SSX
- Chromosome Location
- Chr. X: 52751204 - 52761536 on Build GRCh38
- UniGene ID
- Hs.289105
- Species
- Homo sapiens
- Gene Symbol
- SSX3
- Entrez Gene ID
- 10214
- Gene Name
- SSX family member 3
- Gene Aliases
- CT5.3
- Chromosome Location
- Chr. X: 48346428 - 48356753 on Build GRCh38
- UniGene ID
- Hs.558445
- Species
- Homo sapiens
- Gene Symbol
- SSX4B
- Entrez Gene ID
- 548313
- Gene Name
- SSX family member 4B
- Gene Aliases
- CT5.4
- Chromosome Location
- Chr. X: 48402082 - 48411910 on Build GRCh38
- UniGene ID
- Hs.558402
- Species
- Homo sapiens
- Gene Symbol
- SSX5
- Entrez Gene ID
- 6758
- Gene Name
- SSX family member 5
- Gene Aliases
- Chromosome Location
- Chr. X: 48177914 - 48196798 on Build GRCh38
- UniGene ID
- Hs.166198
- Species
- Homo sapiens
- Gene Symbol
- SSX6
- Entrez Gene ID
- 280657
- Gene Name
- SSX family member 6, pseudogene
- Gene Aliases
- SSX6P, SSXP2, dJ54B20.1, psiSSX2
- Chromosome Location
- Chr. X: 48107982 - 48120686 on Build GRCh38
- UniGene ID
- Hs.511998
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028366.1 |
|
8 |
747 |
|
- Gene Symbol
- SSX7
- Entrez Gene ID
- 280658
- Gene Name
- SSX family member 7
- Gene Aliases
- Chromosome Location
- Chr. X: 52644061 - 52654900 on Build GRCh38
- UniGene ID
- Hs.558712
- Species
- Homo sapiens
View Full Product Details