Gene Transcripts

Gene Symbol
WWOX
Entrez Gene ID
51741
Gene Name
WW domain containing oxidoreductase
Gene Aliases
D16S432E, EIEE28, FOR, FRA16D, HHCMA56, PRO0128, SCAR12, SDR41C1, WOX1
Chromosome Location
Chr. 16: 78099413 - 79212667 on Build GRCh38
UniGene ID
Hs.461453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016373.3 NP_057457.1 2 506
NM_130791.3 NP_570607.1 2 506
NR_120436.1 2 620
XM_011523101.2 XP_011521403.1 2 265
XM_011523103.2 XP_011521405.1 2 265
XM_011523104.2 XP_011521406.1 2 265
XM_011523105.2 XP_011521407.1 2 265
XM_017023278.1 XP_016878767.1 2 265
GenBank mRNA AF187015.1 - 2 224
AF211943.1 - 2 265
AF227526.1 - 2 225
AF227527.1 - 2 244
AF227528.1 - 2 241
AF395123.1 - 2 273
AF395124.1 - 2 273
AK027626.1 - 2 264
AK290438.1 - 2 256
AK291300.1 - 2 263
AW874692.1 - 2 151
AY256821.1 - 2 195
BC003184.1 - 2 219 3535809
BU157392.1 - 2 247
DB035405.1 - 2 506

View Full Product Details