Gene Transcripts

Gene Symbol
WWOX
Entrez Gene ID
51741
Gene Name
WW domain containing oxidoreductase
Gene Aliases
D16S432E, EIEE28, FOR, FRA16D, HHCMA56, PRO0128, SCAR12, SDR41C1, WOX1
Chromosome Location
Chr. 16: 78099413 - 79212667 on Build GRCh38
UniGene ID
Hs.461453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120435.1 1 467
NR_120436.1 1 467
GenBank mRNA AF227529.1 - 1 205
AW874693.1 - 1 446
BU157392.1 - 1 94

View Full Product Details