Gene Transcripts
- Gene Symbol
- CCL15-CCL14
- Entrez Gene ID
- 348249
- Gene Name
- CCL15-CCL14 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 17: 35983656 - 36002038 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027921.3 |
|
1 |
256 |
|
NR_027922.3 |
|
1 |
256 |
|
- Gene Symbol
- CCL14
- Entrez Gene ID
- 6358
- Gene Name
- C-C motif chemokine ligand 14
- Gene Aliases
- CC-1, CC-3, CKB1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, MCIF, NCC-2, NCC2, SCYA14, SCYL2, SY14
- Chromosome Location
- Chr. 17: 35983656 - 35986728 on Build GRCh38
- UniGene ID
- Hs.272493
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM853193.1 |
- |
1 |
302 |
|
- Gene Symbol
- CCL15
- Entrez Gene ID
- 6359
- Gene Name
- C-C motif chemokine ligand 15
- Gene Aliases
- HCC-2, HMRP-2B, LKN-1, LKN1, MIP-1 delta, MIP-1D, MIP-5, MRP-2B, NCC-3, NCC3, SCYA15, SCYL3, SY15
- Chromosome Location
- Chr. 17: 35997582 - 36002038 on Build GRCh38
- UniGene ID
- Hs.272493
- Species
- Homo sapiens
View Full Product Details