Gene Transcripts

Gene Symbol
SLX1B
Entrez Gene ID
79008
Gene Name
SLX1 homolog B, structure-specific endonuclease subunit
Gene Aliases
GIYD2
Chromosome Location
Chr. 16: 29454501 - 29458224 on Build GRCh38
UniGene ID
Hs.728161
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024044.3 NP_076949.1 6 1135
NM_178044.2 NP_835145.1 5 793
GenBank mRNA AK027092.1 - 5, 6 999
AK225436.1 - 5, 6 998
BC000754.2 - 6 1082 2823474
BC000803.1 - 6 929 3452236
BC015990.2 - 6 1088 3503887
BC019306.1 - 6 1082 2823474
BC069007.1 - 5 732 4335620

Gene Symbol
SLX1A
Entrez Gene ID
548593
Gene Name
SLX1 homolog A, structure-specific endonuclease subunit
Gene Aliases
GIYD1
Chromosome Location
Chr. 16: 30193843 - 30197566 on Build GRCh38
UniGene ID
Hs.729791
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014999.2 NP_001014999.1 6 1135
NM_001015000.2 NP_001015000.1 5 793

View Full Product Details