Gene Transcripts

Gene Symbol
FCAR
Entrez Gene ID
2204
Gene Name
Fc fragment of IgA receptor
Gene Aliases
CD89, CTB-61M7.2, FcalphaRI
Chromosome Location
Chr. 19: 54874093 - 54891420 on Build GRCh38
UniGene ID
Hs.659872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002000.3 NP_001991.1 5 1529
NM_133269.3 NP_579803.1 5 1463
NM_133271.3 NP_579805.1 4 1241
NM_133272.3 NP_579806.1 4 1493
NM_133273.3 NP_579807.1 3 1205
NM_133274.3 NP_579808.1 4 1419
NM_133277.3 NP_579811.1 3 1202
NM_133278.3 NP_579812.1 4 1427
XM_011526625.2 XP_011524927.1 5 1453
XM_017026473.1 XP_016881962.1 5 1502
XM_017026474.1 XP_016881963.1 4 1285

View Full Product Details