Gene Transcripts

Gene Symbol
Eny2
Entrez Gene ID
685258
Gene Name
enhancer of yellow 2 homolog (Drosophila)
Gene Aliases
Chromosome Location
Chr. 7: 83348656 - 83358430 on Build Rnor_6.0
UniGene ID
Rn.3365
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130580.1 NP_001124052.1 5 1554
XM_006241615.2 XP_006241677.1 5 1520

View Full Product Details