Gene Transcripts
- Gene Symbol
- RUNX1
- Entrez Gene ID
- 861
- Gene Name
- runt related transcription factor 1
- Gene Aliases
- AML1, AML1-EVI-1, AMLCR1, CBF2alpha, CBFA2, EVI-1, PEBP2aB, PEBP2alpha
- Chromosome Location
- Chr. 21: 34787801 - 35049334 on Build GRCh38
- UniGene ID
- Hs.149261
- Species
- Homo sapiens
- Gene Symbol
- SIN3A
- Entrez Gene ID
- 25942
- Gene Name
- SIN3 transcription regulator family member A
- Gene Aliases
- Chromosome Location
- Chr. 15: 75369379 - 75455819 on Build GRCh38
- UniGene ID
- Hs.513039
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
KT336104.1 |
- |
|
393 |
|
View Full Product Details