Gene Transcripts

Gene Symbol
PLEKHO1
Entrez Gene ID
51177
Gene Name
pleckstrin homology domain containing O1
Gene Aliases
CKIP-1, CKIP1, JBP, OC120
Chromosome Location
Chr. 1: 150149434 - 150160065 on Build GRCh38
UniGene ID
Hs.438824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304722.1 NP_001291651.1 2, 3 501
NM_001304723.1 NP_001291652.1 1, 2 474
NM_001304724.1 NP_001291653.1 2, 3 501
NM_016274.5 NP_057358.2 2, 3 534
XM_017001420.1 XP_016856909.1 2, 3 459
GenBank mRNA AF073836.1 - 1, 2 164
AF168676.1 - 2, 3 266
AF192912.1 - 2, 3 471
AF217956.1 - 2, 3 322
AF291105.1 - 2, 3 462
AK222921.1 - 2, 3 178
AK302255.1 - 2, 3 518
BC010149.2 - 2, 3 262 4330014
BC023533.2 - 2, 3 358
BP329810.1 - 1, 2 223
BX376480.2 - 2, 3 470
DA604331.1 - 2, 3 370
DB023901.1 - 1, 2 474

View Full Product Details