Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018842.4 | NP_061330.2 | 9 | 1145 | |
GenBank mRNA | AF119666.2 | - | 9 | 1098 | |
AK027109.1 | - | 5 | 600 | ||
AK309936.1 | - | 9 | 1120 | ||
BC013888.2 | - | 9 | 1098 | 3842949 |