Gene Transcripts

Gene Symbol
ZNF837
Entrez Gene ID
116412
Gene Name
zinc finger protein 837
Gene Aliases
Chromosome Location
Chr. 19: 58367623 - 58381022 on Build GRCh38
UniGene ID
Hs.222236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138466.1 NP_612475.1 3 1907
NR_049780.1 3 2009
XM_017026242.1 XP_016881731.1 2 2675
GenBank mRNA BC012365.1 - 2 1769

View Full Product Details