Gene Transcripts

Gene Symbol
SNX32
Entrez Gene ID
254122
Gene Name
sorting nexin 32
Gene Aliases
SNX6B
Chromosome Location
Chr. 11: 65833636 - 65854687 on Build GRCh38
UniGene ID
Hs.591950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152760.2 NP_689973.2 13 1298
XM_017017470.1 XP_016872959.1 13 1576
XM_017017471.1 XP_016872960.1 14 1760
XM_017017472.1 XP_016872961.1 12 1962
GenBank mRNA AK289587.1 - 13 1264
AL833261.1 - 13 1284
BC040981.2 - 13 1274 4797976
BC045563.1 - 13 1247 4815203

View Full Product Details