Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000520.5 | NP_000511.2 | 7 | 1229 | |
NM_001318825.1 | NP_001305754.1 | 7 | 1262 | ||
NR_134869.1 | 7 | 1229 | |||
GenBank mRNA | AK222502.1 | - | 7 | 742 | |
AK296528.1 | - | 7 | 600 | ||
AK297678.1 | - | 6 | 661 | ||
AK301000.1 | - | 7 | 782 | ||
AK301138.1 | - | 5 | 601 | ||
AK307770.1 | - | 7 | 749 | ||
AK310154.1 | - | 6 | 712 | ||
BC001138.2 | - | 5 | 369 | ||
BC018927.2 | - | 7 | 739 | 3353424 | |
BC021030.1 | - | 1 | 68 | ||
BC084537.1 | - | 7 | 735 | 5459166 | |
BX537989.1 | - | 6 | 6055 | ||
CR627386.1 | - | 7 | 1229 | ||
KU178109.1 | - | 7 | 728 | ||
M13520.1 | - | 7 | 612 |