Gene Transcripts
- Gene Symbol
- HNF1A
- Entrez Gene ID
- 6927
- Gene Name
- HNF1 homeobox A
- Gene Aliases
- HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1
- Chromosome Location
- Chr. 12: 120978515 - 121002512 on Build GRCh38
- UniGene ID
- Hs.654455
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000545.6 |
NP_000536.5
|
3, 4 |
911 |
|
NM_001306179.1 |
NP_001293108.1
|
3, 4 |
911 |
|
XM_005253931.3 |
XP_005253988.1
|
3, 4 |
964 |
|
GenBank mRNA |
AK298422.1 |
- |
3, 4 |
785 |
|
BC104908.1 |
- |
3, 4 |
730 |
8143911 |
BC104910.1 |
- |
3, 4 |
730 |
8143913 |
BC143483.1 |
- |
3, 4 |
730 |
|
HM116541.1 |
- |
3, 4 |
853 |
|
HM116542.1 |
- |
3, 4 |
853 |
|
HM116547.1 |
- |
3, 4 |
853 |
|
HM116548.1 |
- |
3, 4 |
853 |
|
HM116552.1 |
- |
4, 5 |
470 |
|
HM449088.1 |
- |
3, 4 |
733 |
|
HM449089.1 |
- |
2, 3 |
533 |
|
M57732.1 |
- |
3, 4 |
733 |
|
X71346.1 |
- |
3, 4 |
906 |
|
X71347.1 |
- |
3, 4 |
841 |
|
View Full Product Details