Gene Transcripts

Gene Symbol
FCN3
Entrez Gene ID
8547
Gene Name
ficolin 3
Gene Aliases
FCNH, HAKA1
Chromosome Location
Chr. 1: 27369110 - 27374824 on Build GRCh38
UniGene ID
Hs.333383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK075140.1 - 7 739
AY358081.1 - 7 728
AY358283.1 - 7 765
BC020731.1 - 7 739 4692272
CR456808.1 - 8 766

View Full Product Details