Gene Transcripts
- Gene Symbol
- CNTNAP3
- Entrez Gene ID
- 79937
- Gene Name
- contactin associated protein-like 3
- Gene Aliases
- CASPR3, CNTNAP3A
- Chromosome Location
- Chr. 9: 39072767 - 39288303 on Build GRCh38
- UniGene ID
- Hs.128474
- Species
- Homo sapiens
- Gene Symbol
- CNTNAP3B
- Entrez Gene ID
- 728577
- Gene Name
- contactin associated protein-like 3B
- Gene Aliases
- Chromosome Location
- Chr. 9: 41891887 - 42129504 on Build GRCh38
- UniGene ID
- Hs.521495
- Species
- Homo sapiens
- Gene Symbol
- CNTNAP3P2
- Entrez Gene ID
- 643827
- Gene Name
- contactin associated protein-like 3 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 9: 67059460 - 67297156 on Build GRCh38
- UniGene ID
- Hs.521495
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_111893.2 |
|
19 |
3181 |
|
View Full Product Details