Gene Transcripts
- Gene Symbol
- GAS8
- Entrez Gene ID
- 2622
- Gene Name
- growth arrest specific 8
- Gene Aliases
- CILD33, DRC4, GAS11
- Chromosome Location
- Chr. 16: 90019629 - 90044971 on Build GRCh38
- UniGene ID
- Hs.431792
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286205.1 |
NP_001273134.1
|
3, 4 |
534 |
|
NM_001286208.1 |
NP_001273137.1
|
3, 4 |
534 |
|
NM_001286209.1 |
NP_001273138.1
|
3, 4 |
426 |
|
NM_001481.2 |
NP_001472.1
|
3, 4 |
411 |
|
XM_005256304.4 |
XP_005256361.1
|
4, 5 |
1066 |
|
XM_005256309.4 |
XP_005256366.1
|
5, 6 |
791 |
|
XM_006721175.3 |
XP_006721238.1
|
3, 4 |
464 |
|
XM_011522990.2 |
XP_011521292.1
|
3, 4 |
485 |
|
XM_011522991.1 |
XP_011521293.1
|
4, 5 |
640 |
|
XM_011522992.2 |
XP_011521294.1
|
2, 3 |
433 |
|
XM_017023122.1 |
XP_016878611.1
|
5, 6 |
791 |
|
XM_017023123.1 |
XP_016878612.1
|
3, 4 |
490 |
|
XM_017023124.1 |
XP_016878613.1
|
3, 4 |
332 |
|
XM_017023125.1 |
XP_016878614.1
|
3, 4 |
464 |
|
GenBank mRNA |
AF050079.1 |
- |
3, 4 |
410 |
|
AK293958.1 |
- |
3, 4 |
441 |
|
AK294050.1 |
- |
2, 3 |
260 |
|
AK297857.1 |
- |
3, 4 |
394 |
|
AK303505.1 |
- |
3, 4 |
467 |
|
AK304731.1 |
- |
3, 4 |
328 |
|
AK315261.1 |
- |
3, 4 |
359 |
|
BC104785.1 |
- |
3, 4 |
396 |
8143788 |
BC112121.1 |
- |
3, 4 |
396 |
8327589 |
BX647947.1 |
- |
3, 4 |
452 |
|
CR749499.1 |
- |
2, 3 |
444 |
|
DC370728.1 |
- |
3, 4 |
412 |
|
DC371501.1 |
- |
3, 4 |
469 |
|
View Full Product Details