Gene Transcripts
- Gene Symbol
- GPM6B
- Entrez Gene ID
- 2824
- Gene Name
- glycoprotein M6B
- Gene Aliases
- M6B
- Chromosome Location
- Chr. X: 13770922 - 13938824 on Build GRCh38
- UniGene ID
- Hs.495710
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001994.2 |
NP_001001994.1
|
5, 6 |
913 |
|
NM_001001995.2 |
NP_001001995.1
|
6, 7 |
1231 |
|
NM_001001996.2 |
NP_001001996.1
|
6, 7 |
1231 |
|
NM_001318729.1 |
NP_001305658.1
|
5, 6 |
913 |
|
NM_005278.4 |
NP_005269.1
|
5, 6 |
1111 |
|
XM_005274489.4 |
XP_005274546.1
|
5, 6 |
1110 |
|
XM_011545497.2 |
XP_011543799.1
|
6, 7 |
922 |
|
XM_017029432.1 |
XP_016884921.1
|
6, 7 |
921 |
|
XM_017029433.1 |
XP_016884922.1
|
5, 6 |
615 |
|
XM_017029434.1 |
XP_016884923.1
|
5, 6 |
638 |
|
GenBank mRNA |
AB209525.1 |
- |
5, 6 |
632 |
|
AK095657.1 |
- |
6, 7 |
985 |
|
AK294314.1 |
- |
5, 6 |
684 |
|
AK299675.1 |
- |
7, 8 |
979 |
|
BC008151.1 |
- |
5, 6 |
737 |
4215416 |
BC047295.1 |
- |
6, 7 |
941 |
4215095 |
BG532134.1 |
- |
1, 2 |
33 |
|
U45955.1 |
- |
5, 6 |
711 |
|
View Full Product Details